Assay Details
Target Gene Details
Entrez Gene ID: | 55634 |
Gene Name: | KRAB box domain containing 4 |
Gene Aliases: |
ZNF673 |
Location: |
Chr.X:46447189-46474639 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRBOX4 | NM_001129898.1 | NP_001123370.1 | ||
| NM_001129899.1 | NP_001123371.1 | |||
| NM_001129900.1 | NP_001123372.1 | |||
| NM_017776.2 | NP_060246.2 | |||
| AK000351.1 | BAA91104.1 | |||
| AK096413.1 | ||||
| AK289703.1 | ||||
| BC012569.1 | AAH12569.1 | |||
| DA152034.1 | ||||
| DA631861.1 | ||||
| DA877282.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576854 | Chr.X:46457004 - 46479835 on Build GRCh38 | Gain |
|
| nsv526411 | Chr.X:45810991 - 46476384 on Build GRCh38 | Gain |
|
| esv3576853 | Chr.X:46169680 - 46644654 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map