Assay Details
Target Gene Details
Entrez Gene ID: | 7224 |
Gene Name: | transient receptor potential cation channel subfamily C member 5 |
Gene Aliases: |
PPP1R159, TRP5 |
Location: |
Chr.X:111774314-112082943 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRPC5 | NM_012471.2 | NP_036603.1 | ||
| XM_017029774.1 | XP_016885263.1 | |||
| AB209258.1 | BAD92495.1 | |||
| AF054568.1 | AAF00002.1 | |||
| BC137271.1 | ||||
| BC137274.1 |
More Information
Set Membership: |
|

Genomic Map