Assay Details
Target Gene Details
Entrez Gene ID: | 9758 |
Gene Name: | FERM and PDZ domain containing 4 |
Gene Aliases: |
PDZD10, PDZK10 |
Location: |
Chr.X:12137329-12724523 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FRMPD4 | NM_014728.3 | NP_055543.2 | ||
XM_011545613.2 | XP_011543915.1 | |||
XM_017029983.1 | XP_016885472.1 | |||
XM_017029986.1 | XP_016885475.1 | |||
AB002314.3 | ||||
AK289694.1 | ||||
BC113700.1 | ||||
BC113702.1 | ||||
BC143746.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
