Assay Details
Target Gene Details
Entrez Gene ID: | 56474 |
Gene Name: | CTP synthase 2 |
Gene Aliases: |
- |
Location: |
Chr.X:16587999-16712979 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTPS2 | NM_001144002.1 | NP_001137474.1 | ||
| NM_019857.4 | NP_062831.3 | |||
| NM_175859.2 | NP_787055.1 | |||
| XM_005274562.3 | XP_005274619.1 | |||
| XM_006724503.3 | XP_006724566.1 | |||
| XM_011545545.2 | XP_011543847.1 | |||
| AF086422.1 | ||||
| AF226667.1 | AAF91241.1 | |||
| AK023549.1 | BAB14607.1 | |||
| AK024070.1 | BAB14814.1 | |||
| AK025654.1 | BAB15204.1 | |||
| AK125332.1 | ||||
| AK125348.1 | ||||
| AK309508.1 | ||||
| BC006256.2 | AAH06256.2 | |||
| BC034986.1 | AAH34986.1 |
More Information
Set Membership: |
|

Genomic Map