Assay Details
Target Gene Details
Entrez Gene ID: | 546 |
Gene Name: | ATRX, chromatin remodeler |
Gene Aliases: |
ATR2, JMS, MRX52, MRXHF1, RAD54, RAD54L, SFM1, SHS, XH2, XNP, ZNF-HX |
Location: |
Chr.X:77504878-77786269 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
More Information
Set Membership: |
|

Genomic Map