Assay Details
Target Gene Details
Entrez Gene ID: | 5251 |
Gene Name: | phosphate regulating endopeptidase homolog, X-linked |
Gene Aliases: |
HPDR, HPDR1, HYP, HYP1, LXHR, PEX, XLH |
Location: |
Chr.X:22032327-22251310 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PHEX | NM_000444.5 | NP_000435.3 | ||
NM_001282754.1 | NP_001269683.1 | |||
XM_011545533.1 | XP_011543835.1 | |||
XM_011545536.2 | XP_011543838.1 | |||
XM_017029579.1 | XP_016885068.1 | |||
AD000712.1 | AAB51604.1 | |||
AK298030.1 | ||||
AK301528.1 | ||||
AK304551.1 | ||||
BC105057.1 | AAI05058.1 | |||
BC105059.1 | AAI05060.1 | |||
U60475.1 | AAC50552.1 | |||
U75645.1 | AAB47749.1 | |||
U82970.1 | AAC24487.1 | |||
U87284.1 | AAB47562.1 |
Target Gene Details
Entrez Gene ID: | 100873942 |
Gene Name: | PHEX antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.X:22162732-22172983 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PHEX-AS1 | NR_046639.1 | |||
AK095185.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
