Assay Details
Target Gene Details
Entrez Gene ID: | 1730 |
Gene Name: | diaphanous related formin 2 |
Gene Aliases: |
DIA, DIA2, DRF2, POF, POF2 |
Location: |
Chr.X:96684663-97600598 on Build GRCh38 |
Assay Gene Location: | Within Intron 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DIAPH2 | NM_006729.4 | NP_006720.1 | ||
| U69179.1 | ||||
| Y15909.1 | CAA75870.1 |
Target Gene Details
Entrez Gene ID: | 10824 |
Gene Name: | DIAPH2 antisense RNA 1 |
Gene Aliases: |
EPAG |
Location: |
Chr.X:97528364-97564535 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DIAPH2-AS1 | NR_125391.1 | |||
| AB593036.1 | ||||
| AB593037.1 |
More Information
Set Membership: |
|

Genomic Map