Assay Details
Target Gene Details
Entrez Gene ID: | 169966 |
Gene Name: | family with sequence similarity 46 member D |
Gene Aliases: |
CT1.26, CT112 |
Location: |
Chr.X:80335504-80445311 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM46D | NM_001170574.1 | NP_001164045.1 | ||
| AK128147.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6972 | Chr.X:80370133 - 80399302 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map