Assay Details
Target Gene Details
Entrez Gene ID: | 54960 |
Gene Name: | gem nuclear organelle associated protein 8 |
Gene Aliases: |
FAM51A1 |
Location: |
Chr.X:14002494-14030087 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GEMIN8 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528634 | Chr.X:13696631 - 14038859 on Build GRCh38 | Gain |
|

Genomic Map