Assay Details
Target Gene Details
Entrez Gene ID: | 51442 |
Gene Name: | vestigial like family member 1 |
Gene Aliases: |
TDU, VGL1 |
Location: |
Chr.X:136532152-136556807 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VGLL1 | NM_016267.3 | NP_057351.1 | ||
| AF137387.1 | AAF13149.1 | |||
| AK309912.1 | ||||
| BC000045.1 | AAH00045.1 | |||
| BC003362.1 | AAH03362.1 |
More Information
Set Membership: |
|

Genomic Map