Assay Details
Target Gene Details
Entrez Gene ID: | 9754 |
Gene Name: | StAR related lipid transfer domain containing 8 |
Gene Aliases: |
ARHGAP38, DLC3, STARTGAP3 |
Location: |
Chr.X:68647669-68725842 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STARD8 | NM_001142503.2 | NP_001135975.1 | ||
| NM_014725.4 | NP_055540.2 | |||
| XM_005262314.4 | XP_005262371.1 | |||
| BC035587.1 | AAH35587.1 | |||
| CR749411.1 | CAH18253.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516469 | Chr.X:68640253 - 68949573 on Build GRCh38 | Gain |
|
| nsv6943 | Chr.X:68637870 - 68674421 on Build GRCh38 | Insertion |
|
| esv2669195 | Chr.X:68603983 - 68737974 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115321491] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map