Assay Details
Target Gene Details
Entrez Gene ID: | 2556 |
Gene Name: | gamma-aminobutyric acid type A receptor alpha3 subunit |
Gene Aliases: |
- |
Location: |
Chr.X:152166234-152451359 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GABRA3 | NM_000808.3 | NP_000799.1 | ||
XM_006724811.2 | XP_006724874.1 | |||
A28102.1 | CAA01919.1 | |||
AK312404.1 | ||||
BC028629.1 | AAH28629.1 | |||
KJ620007.1 | ||||
S62908.1 | AAB27279.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv516934 | Chr.X:152163297 - 152511752 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
