Assay Details
Target Gene Details
Entrez Gene ID: | 4128 |
Gene Name: | monoamine oxidase A |
Gene Aliases: |
MAO-A |
Location: |
Chr.X:43654907-43746824 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAOA | NM_000240.3 | 1 | 824 | NP_000231.1 |
| NM_001270458.1 | 1 | 824 | NP_001257387.1 | |
| BC044787.1 | 1 | 826 | AAH44787.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517873 | Chr.X:43545579 - 43678961 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map