Assay Details
Target Gene Details
Entrez Gene ID: | 25975 |
Gene Name: | EGF like domain multiple 6 |
Gene Aliases: |
MAEG, W80 |
Location: |
Chr.X:13569575-13633575 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EGFL6 | NM_001167890.1 | NP_001161362.1 | ||
| NM_015507.3 | NP_056322.2 | |||
| AF186084.1 | AAF27812.1 | |||
| AF193055.1 | AAG22483.1 | |||
| AJ245671.1 | CAB92132.1 | |||
| AK075214.1 | BAC11477.1 | |||
| AK300137.1 | ||||
| AK300236.1 | ||||
| AK315018.1 | ||||
| AL117610.1 | CAB56014.2 | |||
| AY358333.1 | AAQ88699.1 | |||
| BC038587.1 | AAH38587.1 |
More Information
Set Membership: |
|

Genomic Map