Assay Details
Target Gene Details
Entrez Gene ID: | 4435 |
Gene Name: | Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 1 |
Gene Aliases: |
MSG1 |
Location: |
Chr.X:72301638-72307187 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CITED1 | NM_001144885.1 | NP_001138357.1 | ||
| NM_001144886.1 | NP_001138358.1 | |||
| NM_001144887.1 | NP_001138359.1 | |||
| NM_004143.3 | NP_004134.2 | |||
| XM_011530958.1 | XP_011529260.1 | |||
| BC004240.1 | AAH04240.1 | |||
| BP247232.1 | ||||
| N27803.1 | ||||
| U65092.1 | AAC51113.1 |
More Information
Set Membership: |
|

Genomic Map