Assay Details
Target Gene Details
Entrez Gene ID: | 55787 |
Gene Name: | taxilin gamma |
Gene Aliases: |
CXorf15, ELRG, FIAT, LSR5, TXLNGX |
Location: |
Chr.X:16786432-16844519 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TXLNG | NM_001168683.1 | NP_001162154.1 | ||
| NM_018360.2 | NP_060830.2 | |||
| AK002071.1 | BAA92068.1 | |||
| AY739713.1 | AAW65982.1 | |||
| BC101572.1 | AAI01573.1 | |||
| BC101576.1 | AAI01577.1 | |||
| DB098819.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2668322 | Chr.X:16781822 - 16787969 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map