Assay Details
Target Gene Details
Entrez Gene ID: | 8409 |
Gene Name: | ubiquitously expressed prefoldin like chaperone |
Gene Aliases: |
ART-27, STAP1 |
Location: |
Chr.X:47651792-47659180 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UXT | NM_004182.3 | NP_004173.1 | ||
| NM_153477.2 | NP_705582.1 | |||
| NR_045559.1 | ||||
| NR_045560.1 | ||||
| AF083241.1 | AAD39839.1 | |||
| AF083242.1 | AAD39840.1 | |||
| AF092737.1 | AAD28698.1 | |||
| AK223320.1 | ||||
| AK312072.1 | ||||
| BC000720.1 | AAH00720.1 | |||
| BC008890.1 | AAH08890.1 | |||
| BM471483.1 | ||||
| DA372561.1 | ||||
| EL949569.1 |
More Information
Set Membership: |
|

Genomic Map