Assay Details
Target Gene Details
Entrez Gene ID: | 9411 |
Gene Name: | Rho GTPase activating protein 29 |
Gene Aliases: |
PARG1 |
Location: |
Chr.1:94168907-94275068 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP29 | NM_004815.3 | 23 | 6587 | NP_004806.3 |
| AK092668.1 | 1 | 816 | ||
| AK310491.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs116611244] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map