Assay Details
Target Gene Details
Entrez Gene ID: | 388662 |
Gene Name: | solute carrier family 6 member 17 |
Gene Aliases: |
MRT48, NTT4 |
Location: |
Chr.1:110150510-110202202 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC6A17 | NM_001010898.2 | NP_001010898.1 | ||
XM_006710643.2 | XP_006710706.1 | |||
AK289982.1 | ||||
DA081361.1 | ||||
DA325382.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv2354 | Chr.1:110148781 - 110180477 on Build GRCh38 | Insertion |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
