Assay Details
Target Gene Details
Entrez Gene ID: | 3192 |
Gene Name: | heterogeneous nuclear ribonucleoprotein U |
Gene Aliases: |
HNRNPU-AS1, HNRPU, SAF-A, SAFA, U21.1, hnRNP U |
Location: |
Chr.1:244842123-244864720 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HNRNPU | XM_017001115.1 | 14 | 8209 | XP_016856604.1 |
| XM_017001116.1 | 14 | 8014 | XP_016856605.1 | |
| XM_017001117.1 | 14 | 8003 | XP_016856606.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv549493 | Chr.1:244721983 - 244856983 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map