Assay Details
Target Gene Details
Entrez Gene ID: | 80135 | 
Gene Name: | ribosome production factor 1 homolog | 
Gene Aliases: | 
	
		BXDC5 | 
Location: | 
		
			Chr.1:84479237-84498350 on Build GRCh38 | 
		Assay Gene Location: | Within Intron 3 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| RPF1 | NM_025065.6 | NP_079341.2 | ||
| AF322053.1 | AAL37236.1 | |||
| AK022537.1 | BAB14086.1 | |||
| AL359584.2 | CAH10671.1 | |||
| BC016051.1 | AAH16051.1 | |||
| CR627468.1 | CAH10670.1 | 
Target Copy Number Variation Details
	    DGV Version: |  
	    Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation  |  
		            Location | CNV Subtype  |  
		            Genes | 
|---|---|---|---|
| nsv830437 | Chr.1:84371841 - 84535586 on Build GRCh38 | Gain+Loss | 
							
								 | 
					
More Information
Set Membership: | 
		   
					
						
							 | 
				

Genomic Map