Assay Details
Target Gene Details
Entrez Gene ID: | 22823 |
Gene Name: | metal response element binding transcription factor 2 |
Gene Aliases: |
M96, PCL2, TDRD19A, dJ976O13.2 |
Location: |
Chr.1:93079219-93139081 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MTF2 | NM_001164391.1 | NP_001157863.1 | ||
NM_001164392.1 | NP_001157864.1 | |||
NM_001164393.1 | NP_001157865.1 | |||
NM_007358.3 | NP_031384.1 | |||
XM_011541016.1 | XP_011539318.1 | |||
XM_011541017.2 | XP_011539319.1 | |||
XM_011541019.1 | XP_011539321.1 | |||
XM_017000676.1 | XP_016856165.1 | |||
XM_017000677.1 | XP_016856166.1 | |||
XM_017000678.1 | XP_016856167.1 | |||
XM_017000679.1 | XP_016856168.1 | |||
AF086908.1 | AAP97138.1 | |||
AJ010014.1 | CAA08970.1 | |||
AK290318.1 | ||||
AK302776.1 | ||||
AK302900.1 | ||||
BC010013.2 | AAH10013.1 | |||
DB173315.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv508404 | Chr.1:93081826 - 93114455 on Build GRCh38 | Deletion |
![]() |
nsv1008602 | Chr.1:92711254 - 93175251 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv830614 | Chr.1:93000421 - 93213143 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv1866 | Chr.1:93095373 - 93127336 on Build GRCh38 | Insertion |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
