Assay Details
Target Gene Details
| Entrez Gene ID: | 23421 | 
| Gene Name: | integrin subunit beta 3 binding protein | 
| Gene Aliases: | CENP-R, CENPR, HSU37139, NRIF3, TAP20 | 
| Location: | Chr.1:63440770-63523517 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 6 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| ITGB3BP | NM_001206739.1 | NP_001193668.1 | ||
| NM_014288.4 | NP_055103.3 | |||
| NR_045147.1 | ||||
| XM_006710514.3 | XP_006710577.3 | |||
| XM_006710515.3 | XP_006710578.3 | |||
| XM_017000853.1 | XP_016856342.1 | |||
| XM_017000854.1 | XP_016856343.1 | |||
| XM_017000855.1 | XP_016856344.1 | |||
| XM_017000857.1 | XP_016856346.1 | |||
| AF175306.1 | AAF00239.1 | |||
| AK309622.1 | ||||
| AK312944.1 | ||||
| BC009929.2 | AAH09929.1 | |||
| BC014385.1 | AAH14385.1 | |||
| BU853146.1 | ||||
| U37139.1 | AAC50294.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv546434 | Chr.1:63449672 - 63576116 on Build GRCh38 | Loss |  EFCAB7  DLEU2L  ITGB3BP | 
| nsv470714 | Chr.1:63301926 - 63537185 on Build GRCh38 | Gain |  FOXD3-AS1  MIR6068  ALG6  EFCAB7  FOXD3  LINC00466  ITGB3BP | 
| nsv546433 | Chr.1:63288065 - 63537185 on Build GRCh38 | Gain |  FOXD3-AS1  MIR6068  ALG6  EFCAB7  FOXD3  LINC00466  ITGB3BP | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top