Assay Details
Target Gene Details
Entrez Gene ID: | 5586 |
Gene Name: | protein kinase N2 |
Gene Aliases: |
PAK2, PRK2, PRKCL2, PRO2042, Pak-2, STK7 |
Location: |
Chr.1:88684144-88836255 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PKN2 | NM_001320707.1 | NP_001307636.1 | ||
NM_001320708.1 | NP_001307637.1 | |||
NM_001320709.1 | NP_001307638.1 | |||
NM_006256.3 | NP_006247.1 | |||
XM_017001782.1 | XP_016857271.1 | |||
XM_017001783.1 | XP_016857272.1 | |||
AF118089.2 | AAF22033.1 | |||
AK298595.1 | ||||
AK300304.1 | ||||
BC125199.1 | ||||
DA451427.1 | ||||
S75548.1 | AAB33346.1 | |||
U33052.1 | AAC50208.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
