Assay Details
Target Gene Details
Entrez Gene ID: | 4259 |
Gene Name: | microsomal glutathione S-transferase 3 |
Gene Aliases: |
GST-III |
Location: |
Chr.1:165630873-165656136 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MGST3 | NM_004528.3 | NP_004519.1 | ||
XM_005245174.3 | XP_005245231.2 | |||
AK312103.1 | ||||
BC000505.2 | AAH00505.1 | |||
BC005964.1 | AAH05964.1 | |||
BI666647.1 | ||||
DQ185045.1 | ABD14425.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
