Assay Details
Target Gene Details
Entrez Gene ID: | 2707 |
Gene Name: | gap junction protein beta 3 |
Gene Aliases: |
CX31, DFNA2, DFNA2B, EKV |
Location: |
Chr.1:34781189-34786366 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GJB3 | NM_001005752.1 | NP_001005752.1 | ||
| NM_024009.2 | NP_076872.1 | |||
| AF052692.1 | 1 | 146 | AAD11816.1 | |
| AK026139.1 | ||||
| AK312890.1 | ||||
| AL547036.3 | ||||
| BC012918.1 | AAH12918.1 | |||
| BC110640.1 | 1 | 205 | AAI10641.1 | |
| BM763034.1 |
More Information
Set Membership: |
|

Genomic Map