Assay Details
Target Gene Details
Entrez Gene ID: | 10657 |
Gene Name: | KH RNA binding domain containing, signal transduction associated 1 |
Gene Aliases: |
Sam68, p62, p68 |
Location: |
Chr.1:32013694-32060859 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KHDRBS1 | NM_001271878.1 | NP_001258807.1 | ||
| NM_006559.2 | NP_006550.1 | |||
| NR_073498.1 | ||||
| NR_073499.1 | ||||
| AK091346.1 | BAC03643.1 | |||
| AK293369.1 | ||||
| AK303218.1 | ||||
| AK312916.1 | ||||
| BC000717.1 | AAH00717.1 | |||
| BC010132.2 | AAH10132.1 | |||
| BC019109.2 | AAH19109.1 | |||
| M88108.1 | AAA59990.1 | |||
| U78971.1 | AAB47504.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834591 | Chr.1:31905853 - 32059864 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map