Assay Details
Target Gene Details
Entrez Gene ID: | 3925 |
Gene Name: | stathmin 1 |
Gene Aliases: |
C1orf215, LAP18, Lag, OP18, PP17, PP19, PR22, SMN |
Location: |
Chr.1:25884186-25906877 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STMN1 | NM_001145454.2 | NP_001138926.1 | ||
| AK056768.1 | BAB71277.1 | |||
| AK303692.1 | ||||
| BC114443.1 | ||||
| BC114502.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv460884 | Chr.1:25778482 - 25891593 on Build GRCh38 | Loss |
|
| nsv834491 | Chr.1:25726411 - 25914117 on Build GRCh38 | Loss |
|
| nsv945838 | Chr.1:25887321 - 25891977 on Build GRCh38 | Duplication |
|
| nsv834502 | Chr.1:25834281 - 25987428 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map