Assay Details
Target Gene Details
Entrez Gene ID: | 127253 |
Gene Name: | tRNA-yW synthesizing protein 3 homolog |
Gene Aliases: |
C1orf171 |
Location: |
Chr.1:74733143-74766677 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TYW3 | NM_001162916.1 | NP_001156388.1 | ||
NM_138467.2 | NP_612476.1 | |||
NR_027962.1 | ||||
XM_006710347.1 | XP_006710410.1 | |||
AK098237.1 | BAC05267.1 | |||
AK294264.1 | ||||
AK299849.1 | ||||
AK311272.1 | ||||
BC009514.1 | AAH09514.1 | |||
BC071765.1 | AAH71765.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830259 | Chr.1:74623493 - 74775378 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv1002115 | Chr.1:72379872 - 74933684 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
