Assay Details
Target Gene Details
Entrez Gene ID: | 51029 |
Gene Name: | desumoylating isopeptidase 2 |
Gene Aliases: |
C1orf121, CGI-146, DESI, DESI1, DeSI-2, FAM152A, PNAS-4, PPPDE1 |
Location: |
Chr.1:244653048-244709034 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DESI2 | NM_001297746.1 | NP_001284675.1 | ||
| NM_016076.4 | NP_057160.2 | |||
| NR_123735.1 | ||||
| NR_123736.1 | ||||
| XM_011544203.2 | XP_011542505.1 | |||
| XM_011544206.2 | XP_011542508.1 | |||
| AF151904.1 | AAD34141.1 | |||
| AF229834.1 | AAF42919.1 | |||
| AK314664.1 | ||||
| BC004485.2 | AAH04485.1 | |||
| BI459676.1 | ||||
| CB854052.1 | ||||
| CN303012.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3578521 | Chr.1:244654938 - 244675197 on Build GRCh38 | Loss |
|
| nsv528017 | Chr.1:244618430 - 244675520 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs113561974] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map