Assay Details
Target Gene Details
Entrez Gene ID: | 7049 |
Gene Name: | transforming growth factor beta receptor 3 |
Gene Aliases: |
BGCAN, betaglycan |
Location: |
Chr.1:91680343-91906002 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGFBR3 | NM_001195683.1 | 17 | 6122 | NP_001182612.1 |
| NM_001195684.1 | 18 | 5958 | NP_001182613.1 | |
| NM_003243.4 | 17 | 6125 | NP_003234.2 | |
| NR_036634.1 | 18 | 6222 | ||
| AI090023.1 | 1 | 351 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2714684 | Chr.1:91095800 - 93049130 on Build GRCh38 | Deletion |
|
| esv2763581 | Chr.1:91603410 - 91738246 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map