Assay Details
Target Gene Details
Entrez Gene ID: | 84976 |
Gene Name: | dispatched RND transporter family member 1 |
Gene Aliases: |
DISPA |
Location: |
Chr.1:222814514-223005995 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DISP1 | XM_011510073.2 | 1 | 261 | XP_011508375.1 |
| XM_011510075.2 | 1 | 261 | XP_011508377.1 | |
| XM_017002611.1 | 1 | 261 | XP_016858100.1 |
More Information
Set Membership: |
|

Genomic Map