Assay Details
Target Gene Details
| Entrez Gene ID: | 23254 | 
| Gene Name: | kazrin, periplakin interacting protein | 
| Gene Aliases: | KAZ | 
| Location: | Chr.1:13893387-15118048 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| KAZN | XM_005245795.4 | XP_005245852.1 | ||
| XM_011541074.2 | XP_011539376.1 | |||
| XM_011541080.2 | XP_011539382.1 | |||
| XM_017000768.1 | XP_016856257.1 | |||
| XM_017000769.1 | XP_016856258.1 | |||
| XM_017000770.1 | XP_016856259.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| esv3585268 | Chr.1:13913847 - 13928580 on Build GRCh38 | Gain |  KAZN | 
| nsv545567 | Chr.1:13558784 - 14026576 on Build GRCh38 | Gain |  PDPN  PRDM2  KAZN | 
 Back To Top
 Back To TopMore Information
Additional Information:
				    	 
					    	
					        	
					            	For this assay, SNP(s) [rs150760470] are located under a primer or probe  sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
					        	
					    	            
				    	
			    	
			    
			| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top