Assay Details
Target Gene Details
Entrez Gene ID: | 257194 |
Gene Name: | neuronal growth regulator 1 |
Gene Aliases: |
DMML2433, IGLON4, KILON, Ntra |
Location: |
Chr.1:71402942-72282850 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEGR1 | NM_173808.2 | NP_776169.2 | ||
| XM_011541200.2 | XP_011539502.1 | |||
| XM_011541201.2 | XP_011539503.1 | |||
| XM_017000961.1 | XP_016856450.1 | |||
| AK092307.1 | BAC03858.1 | |||
| AK295481.1 | ||||
| BC036771.1 | AAH36771.1 | |||
| BX538014.1 | CAD97961.1 |
Target Gene Details
Entrez Gene ID: | 100852409 |
Gene Name: | NEGR1 intronic transcript 1 |
Gene Aliases: |
- |
Location: |
Chr.1:71794232-71837012 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEGR1-IT1 | NR_046218.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1008210 | Chr.1:71633478 - 71908514 on Build GRCh38 | Loss |
|
| esv3893668 | Chr.1:71818523 - 71871230 on Build GRCh38 | Loss |
|
| esv2762150 | Chr.1:71805544 - 71930270 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map