Assay Details
Target Gene Details
Entrez Gene ID: | 127495 |
Gene Name: | leucine rich repeat containing 39 |
Gene Aliases: |
- |
Location: |
Chr.1:100148448-100178273 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LRRC39 | NM_001256385.1 | NP_001243314.1 | ||
NM_001256386.1 | NP_001243315.1 | |||
NM_001256387.1 | NP_001243316.1 | |||
NM_144620.3 | NP_653221.1 | |||
AK096892.1 | ||||
AL833417.1 | ||||
AY358131.1 | AAQ88498.1 | |||
BC009613.1 | AAH09613.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830804 | Chr.1:100036777 - 100210481 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
