Assay Details
Target Gene Details
Entrez Gene ID: | 54874 |
Gene Name: | formin binding protein 1 like |
Gene Aliases: |
C1orf39, TOCA1 |
Location: |
Chr.1:93448067-93569671 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FNBP1L | NM_001024948.2 | NP_001020119.1 | ||
| NM_001164473.2 | NP_001157945.1 | |||
| NM_017737.4 | NP_060207.2 | |||
| XM_011541625.2 | XP_011539927.1 | |||
| XM_017001533.1 | XP_016857022.1 | |||
| AY514449.1 | AAR98814.1 | |||
| DW423801.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs75978584] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map