Assay Details
Target Gene Details
Entrez Gene ID: | 8832 |
Gene Name: | CD84 molecule |
Gene Aliases: |
LY9B, SLAMF5, hCD84, mCD84 |
Location: |
Chr.1:160541094-160579516 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CD84 | NM_001184879.1 | NP_001171808.1 | ||
NM_001184881.1 | NP_001171810.1 | |||
NM_001184882.1 | NP_001171811.1 | |||
NM_003874.3 | NP_003865.1 | |||
XM_011510094.2 | XP_011508396.1 | |||
XM_011510095.2 | XP_011508397.1 | |||
AF054815.1 | AAF21721.1 | |||
AF054816.1 | AAF21722.1 | |||
AF054817.1 | AAF21723.1 | |||
AF054818.1 | AAF21724.1 | |||
AJ223324.1 | CAA11264.1 | |||
AK296290.1 | ||||
AK313496.1 | ||||
BC020063.1 | AAH20063.1 | |||
CR541847.1 | CAG46645.1 | |||
CR933666.1 | CAI45963.1 | |||
U82988.1 | AAB84364.1 | |||
U96627.1 | AAD04232.1 | |||
Y12632.1 | CAA73181.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831703 | Chr.1:160526334 - 160673234 on Build GRCh38 | Gain |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
