Assay Details
Target Gene Details
Entrez Gene ID: | 374946 |
Gene Name: | dorsal inhibitory axon guidance protein |
Gene Aliases: |
AGPA3119, C1orf187, UNQ3119, neucrin |
Location: |
Chr.1:11686635-11721766 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DRAXIN | NM_198545.3 | NP_940947.3 | ||
| XM_011541405.2 | XP_011539707.1 | |||
| XM_017001214.1 | XP_016856703.1 | |||
| AB301919.1 | ||||
| AK075558.1 | BAC11697.1 | |||
| AK311439.1 | ||||
| AY358750.1 | AAQ89110.1 | |||
| BC111064.1 | AAI11065.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832758 | Chr.1:11637026 - 11777087 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74657824] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map