Assay Details
Target Gene Details
Entrez Gene ID: | 4919 |
Gene Name: | receptor tyrosine kinase like orphan receptor 1 |
Gene Aliases: |
NTRKR1, dJ537F10.1 |
Location: |
Chr.1:63774019-64181498 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ROR1 | NM_001083592.1 | NP_001077061.1 | ||
| NM_005012.3 | NP_005003.2 | |||
| XM_011541526.1 | XP_011539828.1 | |||
| XM_017001376.1 | XP_016856865.1 | |||
| XM_017001377.1 | XP_016856866.1 | |||
| AK308088.1 | ||||
| BC080541.1 | AAH80541.1 | |||
| BC128386.1 | ||||
| M97675.1 | AAA60275.1 |
Target Gene Details
Entrez Gene ID: | 101927034 |
Gene Name: | ROR1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:64094442-64113791 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ROR1-AS1 | NR_110665.1 | |||
| AK096291.1 |
More Information
Set Membership: |
|

Genomic Map