Assay Details
Target Gene Details
Entrez Gene ID: | 100287722 |
Gene Name: | AP4B1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:113812612-113901237 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP4B1-AS1 | NR_037864.1 | |||
| NR_125965.1 | ||||
| AK123199.1 | ||||
| DB028504.1 | ||||
| DB074478.1 |
Target Gene Details
Entrez Gene ID: | 440603 |
Gene Name: | BCL2 like 15 |
Gene Aliases: |
Bfk, C1orf178 |
Location: |
Chr.1:113876814-113887547 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCL2L15 | NM_001010922.2 | NP_001010922.1 | ||
| AI821052.1 | ||||
| AK289439.1 | ||||
| AY265861.1 | AAP93855.1 | |||
| AY265862.1 | AAP93856.1 | |||
| AY265863.1 | AAP93857.1 | |||
| AY265864.1 | AAP93858.1 | |||
| BC127719.1 | ||||
| BC127720.1 | ||||
| CR749373.1 | CAH18226.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3587251 | Chr.1:113871703 - 113936287 on Build GRCh38 | Loss |
|
| esv3587240 | Chr.1:113533246 - 114158910 on Build GRCh38 | Loss |
|
| esv23869 | Chr.1:113320330 - 114358495 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map