Assay Details
Target Gene Details
Entrez Gene ID: | 192670 |
Gene Name: | argonaute 4, RISC catalytic component |
Gene Aliases: |
EIF2C4 |
Location: |
Chr.1:35807630-35857890 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGO4 | NM_017629.3 | NP_060099.2 | ||
| XM_005270578.3 | XP_005270635.1 | |||
| XM_005270579.3 | XP_005270636.1 | |||
| XM_017000529.1 | XP_016856018.1 | |||
| AB046787.1 | ||||
| AL831908.1 | ||||
| BC152450.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834680 | Chr.1:35660621 - 35892216 on Build GRCh38 | Gain |
|
| nsv525430 | Chr.1:35776427 - 35860595 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map