Assay Details
Target Gene Details
Entrez Gene ID: | 65979 |
Gene Name: | phosphatase and actin regulator 4 |
Gene Aliases: |
PPP1R124 |
Location: |
Chr.1:28369574-28500369 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHACTR4 | NM_001048183.1 | NP_001041648.1 | ||
| XM_017002159.1 | XP_016857648.1 | |||
| XM_017002161.1 | XP_016857650.1 | |||
| XM_017002162.1 | XP_016857651.1 | |||
| XM_017002164.1 | XP_016857653.1 | |||
| XM_017002165.1 | XP_016857654.1 | |||
| XM_017002166.1 | XP_016857655.1 | |||
| XM_017002167.1 | XP_016857656.1 | |||
| XM_017002168.1 | XP_016857657.1 | |||
| XM_017002169.1 | XP_016857658.1 | |||
| XM_017002171.1 | XP_016857660.1 | |||
| XM_017002173.1 | XP_016857662.1 | |||
| AK025436.1 | BAB15130.1 | |||
| AL705720.1 | ||||
| BC029266.1 | AAH29266.1 | |||
| CR749449.1 | CAH18286.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545916 | Chr.1:28208722 - 28624350 on Build GRCh38 | Gain |
|
| nsv7568 | Chr.1:28381292 - 28412121 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map