Assay Details
Target Gene Details
| Entrez Gene ID: | 5563 | 
| Gene Name: | protein kinase AMP-activated catalytic subunit alpha 2 | 
| Gene Aliases: | AMPK, AMPK2, AMPKa2, PRKAA | 
| Location: | Chr.1:56645317-56715335 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 5 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| PRKAA2 | NM_006252.3 | NP_006243.2 | ||
| XM_017001692.1 | XP_016857181.1 | |||
| XM_017001693.1 | XP_016857182.1 | |||
| XM_017001694.1 | XP_016857183.1 | |||
| AK314011.1 | ||||
| BC069680.1 | AAH69680.1 | |||
| BC069740.1 | AAH69740.1 | |||
| BC069823.1 | AAH69823.1 | |||
| DA710012.1 | ||||
| EF056019.1 | ||||
| FJ200485.1 | ||||
| FJ200486.1 | ||||
| HQ628625.1 | ||||
| U06454.1 | AAA64745.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv520726 | Chr.1:56678313 - 56739993 on Build GRCh38 | Loss |  PRKAA2  C1orf168 | 
| nsv523032 | Chr.1:56651236 - 56783885 on Build GRCh38 | Gain |  PRKAA2  C1orf168 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top