Assay Details
Target Gene Details
Entrez Gene ID: | 192669 |
Gene Name: | argonaute 3, RISC catalytic component |
Gene Aliases: |
EIF2C3 |
Location: |
Chr.1:35925198-36072037 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGO3 | XM_005270575.3 | 1 | 5454 | XP_005270632.1 |
| XM_017000523.1 | 1 | 5454 | XP_016856012.1 |
More Information
Set Membership: |
|

Genomic Map