Assay Details
Target Gene Details
Entrez Gene ID: | 6195 |
Gene Name: | ribosomal protein S6 kinase A1 |
Gene Aliases: |
HU-1, MAPKAPK1A, RSK, RSK1, p90Rsk |
Location: |
Chr.1:26529758-26575029 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RPS6KA1 | NM_002953.3 | NP_002944.2 | ||
XM_005245966.4 | XP_005246023.1 | |||
AK225672.1 | ||||
AK294818.1 | ||||
AK299007.1 | ||||
AK315730.1 | ||||
BC014966.1 | AAH14966.1 | |||
BM836865.1 | ||||
L07597.1 | AAC82497.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv526179 | Chr.1:26534790 - 26579076 on Build GRCh38 | Gain |
![]() ![]() |
nsv545908 | Chr.1:26509300 - 26661535 on Build GRCh38 | Loss |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
