Assay Details
Target Gene Details
Entrez Gene ID: | 1600 |
Gene Name: | DAB1, reelin adaptor protein |
Gene Aliases: |
- |
Location: |
Chr.1:56997906-58250539 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DAB1 | NM_021080.3 | NP_066566.3 | ||
| AB210012.1 | ||||
| AF071062.1 | AAC70068.1 | |||
| AK095513.1 | ||||
| AK313144.1 | ||||
| BC067445.1 | AAH67445.1 | |||
| BC067446.1 | AAH67446.1 | |||
| BC067447.1 | AAH67447.1 | |||
| BC112938.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546305 | Chr.1:56997653 - 57350817 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map