Assay Details
Target Gene Details
Entrez Gene ID: | 101410535 |
Gene Name: | LAMTOR5 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:110407809-110416274 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LAMTOR5-AS1 | NR_102697.1 | |||
BE547292.1 | ||||
BG718030.1 | ||||
HY022359.1 |
