Assay Details
Target Gene Details
Entrez Gene ID: | 1385 |
Gene Name: | cAMP responsive element binding protein 1 |
Gene Aliases: |
CREB, CREB-1 |
Location: |
Chr.2:207529892-207605989 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CREB1 | NM_004379.4 | 8 | 8117 | NP_004370.1 |
| NM_134442.4 | 9 | 8159 | NP_604391.1 | |
| NR_135473.1 | 10 | 6563 | ||
| AK091534.1 | 1 | 1323 | ||
| AK095116.1 | 1 | 507 |
Target Gene Details
Entrez Gene ID: | 151194 |
Gene Name: | methyltransferase like 21A |
Gene Aliases: |
FAM119A, HCA557b, HSPA-KMT |
Location: |
Chr.2:207580631-207625928 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| METTL21A | NM_001308021.1 | NP_001294950.1 | ||
| XM_011510727.2 | XP_011509029.1 | |||
| XM_011510728.2 | XP_011509030.1 | |||
| XM_011510729.2 | XP_011509031.1 | |||
| XM_017003442.1 | XP_016858931.1 | |||
| XM_017003443.1 | XP_016858932.1 | |||
| XM_017003445.1 | XP_016858934.1 | |||
| XM_017003448.1 | XP_016858937.1 | |||
| XM_017003449.1 | XP_016858938.1 | |||
| XM_017003450.1 | XP_016858939.1 | |||
| BC009462.1 | AAH09462.1 | |||
| CN367990.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3131 | Chr.2:207578177 - 207624543 on Build GRCh38 | Deletion |
|
| nsv1013908 | Chr.2:207577371 - 207629609 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map