Assay Details
Target Gene Details
Entrez Gene ID: | 54891 |
Gene Name: | INO80 complex subunit D |
Gene Aliases: |
- |
Location: |
Chr.2:205993721-206086182 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INO80D | NM_017759.4 | 11 | 8768 | NP_060229.3 |
| XM_011511369.2 | 11 | 8761 | XP_011509671.1 | |
| XM_011511375.2 | 11 | 8486 | XP_011509677.1 | |
| XM_011511376.2 | 11 | 8473 | XP_011509678.1 | |
| CR933609.1 | 1 | 3402 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508196 | Chr.2:205958585 - 206020859 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map