Assay Details
Target Gene Details
Entrez Gene ID: | 100616208 |
Gene Name: | microRNA 4782 |
Gene Aliases: |
- |
Location: |
Chr.2:113721290-113721368 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR4782 | NR_039943.1 |
Target Gene Details
Entrez Gene ID: | 80255 |
Gene Name: | solute carrier family 35 member F5 |
Gene Aliases: |
- |
Location: |
Chr.2:113702677-113756823 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC35F5 | NM_025181.3 | NP_079457.2 | ||
| NR_104470.1 | ||||
| XM_005263799.3 | XP_005263856.1 | |||
| XM_011511921.2 | XP_011510223.1 | |||
| XM_011511922.2 | XP_011510224.1 | |||
| XM_011511923.2 | XP_011510225.1 | |||
| XM_017005027.1 | XP_016860516.1 | |||
| XM_017005028.1 | XP_016860517.1 | |||
| AF529364.1 | AAQ09598.1 | |||
| AK024666.1 | BAB14954.1 | |||
| AK315719.1 | ||||
| AY358620.1 | AAQ88983.1 | |||
| BC018537.1 | AAH18537.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3592016 | Chr.2:113233328 - 113804636 on Build GRCh38 | Gain |
|
| nsv1001195 | Chr.2:113604426 - 113748753 on Build GRCh38 | Gain |
|
| nsv2882 | Chr.2:113695137 - 113729107 on Build GRCh38 | Insertion |
|
| esv3592028 | Chr.2:113650431 - 113744360 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map